missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine, 99+%, Thermo Scientific Chemicals

Product Code. 10092890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10092890 250 mL Glass bottle 250mL
10434335 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10092890 Supplier Thermo Scientific Acros Supplier No. 219432500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-18-9
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008862
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
Synonym diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine, bis propan-2-yl amine, n-isopropylpropan-2-amine, di-isopropylamine, n-isopropyl-1-amino-2-methylethane, diisopropyl amine
PubChem CID 7912
SMILES CC(C)NC(C)C

Specifications

Melting Point -61.0°C
Density 0.7160g/mL
Boiling Point 84.0°C
Flash Point −17°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C6H15N
Linear Formula [(CH3)2CH]2NH
Refractive Index 1.3910 to 1.3930
Quantity 250 mL
Beilstein 04, 154
Merck Index 15, 3215
Specific Gravity 0.716
Solubility Information Solubility in water: 100g/L (20°C). Other solubilities: soluble in alcohol,soluble in most common organic solvents
IUPAC Name N-propan-2-ylpropan-2-amine
Viscosity 0.4 mPa.s (20°C)
Formula Weight 101.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Diisopropylamine, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Diisopropylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.