missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine, 99+%, Thermo Scientific Chemicals

Product Code. 11364489
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11364489

Brand: Thermo Scientific Alfa Aesar A10280.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

 

Diisopropylamine is used as a precursor to prepare lithium diisopropylamide and sulfenamides, which are used in rubber vulcanization. It is also involved in the synthesis of N,N-Diisopropylethylamine (Hünig's base) obtained by alkylation with diethyl sulfate. Diisopropylammonium bromide is a bromide salt of DIPA used as an inert alternative to barium titanate. It is widely used as solvent as well as an intermediate in the the synthesis of pharmaceuticals, dyes, mineral flotation agents and emulsifiers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropylamine is used as a precursor to prepare lithium diisopropylamide and sulfenamides, which are used in rubber vulcanization. It is also involved in the synthesis of N,N-Diisopropylethylamine (Hünig′s base) obtained by alkylation with diethyl sulfate. Diisopropylammonium bromide is a bromide salt of DIPA used as an inert alternative to barium titanate. It is widely used as solvent as well as an intermediate in the the synthesis of pharmaceuticals, dyes, mineral flotation agents and emulsifiers.

Solubility
Miscible with water.

Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids, strong bases, strong oxidizing agents and plastics.
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-18-9
Melting Point -61°C
Density 0.718 g/mL
Boiling Point 83°C to 84°C
Flash Point -7°C
Assay Percent Range >99%
Molecular Formula C6H15N
Refractive Index 1.392
MDL Number MFCD00008862
Quantity 100 mL
Beilstein 605284
Synonym diisopropylamine, 2-propanamine, n-1-methylethyl, n-1-methylethyl-2-propanamine, dipa, n,n-diisopropylamine
Solubility Information Miscible with water.
InChI Key UAOMVDZJSHZZME-UHFFFAOYSA-N
SMILES CC(C)NC(C)C
IUPAC Name N-propan-2-ylpropan-2-amine
Molecular Weight (g/mol) 101.19
PubChem CID 7912
Formula Weight 101.19
Chemical Name or Material Diisopropylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.