missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylaminomethylidene(diisopropyl)ammonium tetrafluoroborate, 97%, Thermo Scientific™

Product Code. 11995961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
Product Code. Quantity Packaging unitSize
11995961 1 g Glass bottle 1g
1 options
This item is not returnable. View return policy

Product Code. 11995961

Brand: Thermo Scientific Acros 435700010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 280.0°C to 284.0°C
CAS 369405-27-6
Color White to Beige
Physical Form Powder/Solid
Quantity 1 g
Assay Percent Range 97%
Molecular Formula C13H29BF4N2
MDL Number MFCD15144860
Synonym n,n,n',n'-tetraisopropylformamidinium tetrafluoroborate, diisopropylamino methylidene diisopropylazanium tetrafluoroborate, bis-diisopropylamino methylium tetrafluoroborate, n-diisopropylamino methylene-n-diisopropylaminium tetrafluoroborate, n-diisopropylamino methylene-n-isopropylpropan-2-aminium tetrafluoroborate, n,n,n inverted exclamation marka,n inverted exclamation marka-tetraisopropylformamidinium tetrafluoroborate
InChI Key PEKVUVXZBLFOHV-UHFFFAOYSA-N
SMILES F[B-](F)(F)F.CC(C)N(C=[N+](C(C)C)C(C)C)C(C)C
IUPAC Name tetrafluoroboranuide; {[bis(propan-2-yl)amino]methylidene}bis(propan-2-yl)azanium
Molecular Weight (g/mol) 300.19
PubChem CID 11077431
Formula Weight 300.19
Percent Purity 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Diisopropylaminomethylidene(diisopropyl)ammonium tetrafluoroborate
Show More Show Less
compliance-icons
Product Identifier
  • Diisopropylaminomethylidene(diisopropyl)ammonium tetrafluoroborate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.