missing translation for 'onlineSavingsMsg'
Learn More

Diltiazem Hydrochloride, TRC

Product Code. 18072630
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18072630 10 mg 10mg
30008323 50 mg 50mg
18052630 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18072630 Supplier Toronto Research Chemicals Supplier No. TRCD46062010MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33286-22-5
Molecular Weight (g/mol) 450.98
IUPAC Name [(2S,3S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride
SMILES Cl.COc1ccc(cc1)[C@@H]2Sc3ccccc3N(CCN(C)C)C(=O)[C@@H]2OC(=O)C

Specifications

Molecular Formula C22 H26 N2 O4 S . Cl H
Quantity 10 mg
Synonym 1,5-Benzothiazepin-4(5H)-one, 3-(acetyloxy)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-, monohydrochloride, (2S,3S)- (9CI), 1,5-Benzothiazepin-4(5H)-one, 3-(acetyloxy)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-, monohydrochloride, (2S-cis)-, (+)-Diltiazem hydrochloride, (2S,3S)-(+)-cis-Diltiazem hydrochloride, (S,S)-Diltiazem hydrochloride, Adizem, Altiazem, Anginyl, Angipress, Angizem, Bi-Teldiem, Blocalcin 60, Britiazim, Bruzem, CRD 401, Calcicard, Cardizem, Cardizem CD, Cardizem SR, Citizem, Cormax, Deltazen, Dicard, Dilacor, Dilacor XR, Diladel, Dilcardia, Dilgard, Dilpral, Dilrene, Diltiazem hydrochloride, Dilzacard, Dilzene, Herbesser, Kaizem, Kardil, Masdil, RG 83606, Tiazac, Tildiem, d-cis-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(p-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one hydrochloride, d-cis-Diltiazem hydrochloride
InChI Formula InChI=1S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21H,13-14H2,1-4H3;1H/t20-,21+;/m1./s1
Formula Weight 450.138
Percent Purity >95%
Chemical Name or Material Diltiazem Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.