missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Dilute Carbol Fuchsin Stain Concentrate

Product Code. 12986307
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
12986307 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12986307 Supplier Pro-Lab Diagnostics™ Supplier No. PL.8004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acid fast stains for mycobacteria. Pro-Lab Diagnostics™ Dilute Carbol Fuchsin Stain Concentrate is concentrated 10x.

  • Pro-Lab Diagnostics offers an extensive range of ready-to-use and concentrated microbiological staining solutions in a selection of convenient and economical sizes.
  • Manufactured in accordance with all current regulatory requirements
  • CE labelled
  • Strict quality control and filtering techniques prior to release
  • Also available are a range of convenient staining kits for Gram, TB and Cryptosporidium staining

Specifications

Type Carbol Fuchsin
Quantity 100 mL
Certifications/Compliance CE marked (98/78/EC)
Format Liquid
For Use With (Application) Gram’s Staining Method for the Initial Differentiation of Gram Positive and Gram Negative Bacteria
Color Magenta
Composition Ethanol 10 - <25% , Phenol 5 - <10% , Methanol 1 - <2.5% , Basic fuchsin 1 - <2.5%
Storage Requirements Keep at temperature not exceeding 20°C
Stain Type Gram
Volume (Metric) 100 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dilute Carbol Fuchsin Concentrate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash contaminated skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use foam, carbon dioxide, dry powder or water fog to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, phenol, basic fuchsin, methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.