missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DIM-C-pPhOH

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
DIM-C-pPhOH is a Nur77 (NR4A1) antagonist. Inhibits TGF-β induced cell migration of breast cancer cell lines. Promotes ROS/endoplasmic reticulum stress and proapoptotic pathways in pancreatic cancer cell lines. Mimics effects of Nur77 RNAi silencing.
Specifications
Specifications
| Chemical Name or Material | 1,1-Bis(3'-indolyl)-1-(p-hydroxyphenyl)methane |
| CAS | 151358-47-3 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C23H18N2O |
| Formula Weight | Observed MW: 338.408 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction