missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ DIM-C-pPhOH
GREENER_CHOICE

Product Code. p-7116134
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18720979

Brand: Tocris Bioscience™ 6377/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nur77 antagonist

DIM-C-pPhOH is a Nur77 (NR4A1) antagonist. Inhibits TGF-β induced cell migration of breast cancer cell lines. Promotes ROS/endoplasmic reticulum stress and proapoptotic pathways in pancreatic cancer cell lines. Mimics effects of Nur77 RNAi silencing.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1-Bis(3'-indolyl)-1-(p-hydroxyphenyl)methane
CAS 151358-47-3
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C23H18N2O
Formula Weight Observed MW: 338.408
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.