missing translation for 'onlineSavingsMsg'
Learn More

Dimaprit dihydrochloride, Tocris Bioscience™

Product Code. 18065352
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18065352 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18065352 Supplier Tocris Bioscience Supplier No. 0506/100

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Standard H2 selective agonist

Selective H2 agonist with central effects upon systemic administration. Also inhibits nNOS (IC50 = 49 μM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 23256-33-9
Molecular Formula C6H17Cl2N3S
Quantity 100 mg
Synonym dimaprit dihydrochloride, diamaprit-2hcl, dimaprit hydrochloride, dimaprit-2hcl, usaf pd-40, 2-3'-dimethylaminopropylthio pseudourea dihydrochloride, 2-3-dimethylaminopropyl-2-thiopseudourea dihydrochloride, 3-dimethylamino propyl carbamimidothioate dihydrochloride, 3-carbamimidoylsulfanyl propyl dimethylamine dihydrochloride, pseudourea, 2-3-dimethylamino propyl-2-thio-, dihydrochloride
InChI Key DFWCPLGXFMSUCW-UHFFFAOYSA-N
SMILES CN(C)CCCSC(=N)N.Cl.Cl
IUPAC Name 3-(dimethylamino)propyl carbamimidothioate;dihydrochloride
Molecular Weight (g/mol) 234.183
PubChem CID 90045
Formula Weight 234.19
Percent Purity >99%
Chemical Name or Material Dimaprit dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.