missing translation for 'onlineSavingsMsg'
Learn More

Dimedone, Pure, Fisher Chemical

Product Code. 10315800 Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
Packaging:
Plastic powder jar
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 10315800

Brand: Fisher Chemical D/3220/46

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 126-81-8
Melting Point 147°C
Color Yellow
Packaging Plastic powder jar
Molecular Formula C8H12O2
MDL Number MFCD00001588
Quantity 25 g
Synonym dimedone, 5,5-dimethyl-1,3-cyclohexanedione, cyclomethone, medon, methone, dimedon, methon, 1,3-cyclohexanedione, 5,5-dimethyl, 5,5-dimethyldihydroresorcinol, 5,5-dimethylhydroresorcinol
InChI Key BADXJIPKFRBFOT-UHFFFAOYSA-N
SMILES CC1(C)CC(=O)CC(=O)C1
IUPAC Name 5,5-dimethylcyclohexane-1,3-dione
Molecular Weight (g/mol) 140.18
PubChem CID 31358
Formula Weight 140.18g/mol
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.