missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimedone, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Dimedone |
| CAS | 126-81-8 |
| Molecular Formula | C8H12O2 |
| Quantity | 50 g |
| Synonym | 5,5-Dimethyl-1,3-cyclohexanedione, 1,1-Dimethyl-3,5-cyclohexanedione, 1,1-Dimethyl-3,5-diketocyclohexane, 5,5-Dimethyl-1,3-cyclohexandione, 5,5-Dimethyl-1,3-cyclohexanedione, 5,5-Dimethyldihydroresorcinol, 5,5-Dimethylhydroresorcinol, Cyclomethone, Dimedon, Lu 274, Medon, Methon, Methone, NSC 14984, NSC 17544, NSC 242994, NSC 43759 |
| InChI Formula | InChI=1S/C8H12O2/c1-8(2)4-6(9)3-7(10)5-8/h3-5H2,1-2H3 |
| SMILES | CC1(C)CC(=O)CC(=O)C1 |
| IUPAC Name | 5,5-dimethylcyclohexane-1,3-dione |
| Molecular Weight (g/mol) | 140.18 |
| Formula Weight | 140.0837 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction