missing translation for 'onlineSavingsMsg'
Learn More

Dimepranol-d6 5mg/mL in methanol, TRC

Product Code. 30405195
Change view
Click to view available options
Quantity:
1x1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30405195 1x1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30405195 Supplier Toronto Research Chemicals Supplier No. TRCKIT34961X1ML
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimepranol-d6
CAS 97964-89-1
Molecular Formula C5H7D6NO
Quantity 1x1 mL
Synonym (±)-1-[(Dimethyl-d6)amino]-2-propanol, (±)-1-(N,N-Dimethyl-d6)amino]-2-propanol, 1-[(Dimethyl-d6)amino]-2-propanol, 1-Methyl-2-[(dimethyl-d6)amino]ethanol, 2-[(Dimethyl-d6)amino]-1-methyl-1-ethanol, 2-[(Dimethyl-d6)amino]-1-methylethanol, 3-[(Dimethyl-d6)amino]-2-propanol, Bisomer Amine D 700, DL-1-[(Dimethyl-d6)amino]-2-propanol, DMA 2P, (Dimethyl-d6)(2-hydroxypropyl)amine, (Dimethyl-d6)isopropanolamine, N,N-(Dimethyl-d6)-2-hydroxy-1-propanamine, N,N-(Dimethyl-d6)-2-hydroxypropylamine, N,N-(Dimethyl-d6)amino-2-propanol, N,N-(Dimethyl-d6)isopropanolamine, NSC 3163
InChI Formula InChI=1S/C5H13NO/c1-5(7)4-6(2)3/h5,7H,4H2,1-3H3/i2D3,3D3
SMILES OC(CN(C([2H])([2H])[2H])C([2H])([2H])[2H])C
Molecular Weight (g/mol) 109.2
Formula Weight 109.1374
Concentration or Composition (by Analyte or Components) 5 mg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.