missing translation for 'onlineSavingsMsg'
Learn More

Dimethallyl carbonate, 97%

Product Code. 11474347
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11474347 25 g 25g
11464347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474347 Supplier Thermo Scientific Alfa Aesar Supplier No. B20414.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64057-79-0 | C9H14O3 | 170.21 g/mol

It is used as an intermediate in enantioselective tsuji enol silane allylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an intermediate in enantioselective tsuji enol silane allylation reaction.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 64057-79-0
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00008598
InChI Key SKCUYCAPQSWGBP-UHFFFAOYSA-N
Synonym dimethallyl carbonate, bis 2-methylallyl carbonate, bis methallyl carbonate, bis 2-methyl-2-propenyl carbonate, 2-propen-1-ol, 2-methyl-, carbonate 2:1, 2-methyl-2-propen-1-ol carbonate 2:1, 2-propen-1-ol, 2-methyl-, 1,1'-carbonate, carbonic acid, bis 2-methylallyl ester, bis 2-methylprop-2-en-1-yl carbonate, acmc-1biju
PubChem CID 47083
IUPAC Name bis(2-methylprop-2-enyl) carbonate
SMILES CC(=C)COC(=O)OCC(C)=C

Especificaciones

Boiling Point 199°C to 202°C
Quantity 25 g
Formula Weight 170.21
Percent Purity 97%
Chemical Name or Material Dimethallyl carbonate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.