missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl-1,3-acetonedicarboxylate, TRC

Product Code. 30369940
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369940 25 g
30384452 100 g
30373737 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369940 Supplier Toronto Research Chemicals Supplier No. TRCD47067525G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1830-54-2
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
Synonym 3-Oxo-pentanedioic Acid 1,5-Dimethyl Ester, 3-Oxo-glutaric Acid Dimethyl Ester, 3-Oxo-pentanedioic Acid Dimethyl Ester, 1,3-Acetonedicarboxylic Acid Dimethyl Ester, 3-Oxopentanedioic Acid Dimethyl Ester, Acetone Dicarboxylic Acid Dimethyl Ester, Dimethyl 1,3-Acetonedicarboxylate, Dimethyl 2-Oxopropane-1,3-dicarboxylate, Dimethyl 3-Ketoglutarate, Dimethyl 3-Oxo-1,5-pentanedioate, Dimethyl 3-Oxoglutarate, Dimethyl 3-Oxopentanedioate, Dimethyl Acetonedicarboxylate, Dimethyl beta-Ketoglutarate, Dimethyl beta-Oxoglutarate, NSC 4677, beta-Oxoglutaric Acid Dimethyl Ester
IUPAC Name dimethyl 3-oxopentanedioate
SMILES COC(=O)CC(=O)CC(=O)OC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C7H10O5/c1-11-6(9)3-5(8)4-7(10)12-2/h3-4H2,1-2H3
Formula Weight 174.0528
Chemical Name or Material Dimethyl-1,3-acetonedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.