missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, 99+%

Product Code. 11446916
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446916 500 g 500g
11426916 25 g 25g
11436916 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446916 Supplier Thermo Scientific Alfa Aesar Supplier No. B24008.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6289-46-9 | C10H12O6 | 228.20 g/mol

Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate is used in the production of pigments such as pigment red 122 and pigment violet 19. It also can react with 2,4-diamino-phenol to get 2,5-bis-(3-amino-4-hydroxy-phenylamino)-cyclohexa-1,4-diene-1,4-dicarboxylic acid dimethyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate is used in the production of pigments such as pigment red 122 and pigment violet 19. It also can react with 2,4-diamino-phenol to get 2,5-bis-(3-amino-4-hydroxy-phenylamino)-cyclohexa-1,4-diene-1,4-dicarboxylic acid dimethyl ester.

Solubility
Soluble in water 0.0016 g/L.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6289-46-9
Molecular Formula C10H12O6
Molecular Weight (g/mol) 228.20
MDL Number MFCD00001607
InChI Key MHKKFFHWMKEBDW-UHFFFAOYNA-N
Synonym dimethyl succinylsuccinate, dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate, succinosuccinic acid, dimethyl ester, dmss, dimethyl succinylosuccinate, dimethyl 2,5-dioxo-1,4-cyclohexanedicarboxylate, dimethyl succinylo succinate, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, dimethyl ester, 2,5-dioxo-1,4-cyclohexanedicarboxylic acid dimethyl ester, 1,4-cyclohexanedicarboxylic acid, 2,5-dioxo-, 1,4-dimethyl ester
PubChem CID 94866
SMILES COC(=O)C1CC(=O)C(CC1=O)C(=O)OC

Specifications

Melting Point 152°C to 156°C
Quantity 500 g
Solubility Information Soluble in water 0.0016g/L.
IUPAC Name 1,4-dimethyl 2,5-dioxocyclohexane-1,4-dicarboxylate
Formula Weight 228.2
Percent Purity ≥99%
Chemical Name or Material Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.