missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (2-oxopropyl)phosphonate, 95%

Product Code. 10092360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10092360 5 g Glass bottle 5g
10154480 25 g Glass bottle 25g
10175353 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10092360 Supplier Thermo Scientific Acros Supplier No. 177610050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4202-14-6 | C5H11O4P | 166.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4202-14-6
Molecular Formula C5H11O4P
Molecular Weight (g/mol) 166.11
MDL Number MFCD00008769
InChI Key UOWIYNWMROWVDG-UHFFFAOYSA-N
Synonym dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone
PubChem CID 77872
IUPAC Name 1-dimethoxyphosphorylpropan-2-one
SMILES CC(=O)CP(=O)(OC)OC

Specifications

Color Colorless to Brown
Boiling Point 76°C to 79°C (3.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3COCH2P(O)(OCH3)2
Refractive Index 1.4380 to 1.441
Quantity 5 g
Formula Weight 166.11
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Dimethyl (2-oxopropyl)phosphonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.