missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (2-Oxopropyl)phosphonate, TRC

Product Code. 30387212
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30387212 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387212 Supplier Toronto Research Chemicals Supplier No. TRCD4767901G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl (2-Oxopropyl)phosphonate
CAS 4202-14-6
Molecular Formula C5H11O4P
Quantity 1 g
Synonym (2-Oxopropyl)phosphonic Acid Dimethyl Ester, (Dimethylphosphono)acetone
InChI Formula InChI=1S/C5H11O4P/c1-5(6)4-10(7,8-2)9-3/h4H2,1-3H3
SMILES O=C(C)CP(OC)(OC)=O
IUPAC Name dimethyl (2-oxopropyl)phosphonate
Molecular Weight (g/mol) 166.11
Formula Weight 166.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.