missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate, 97%

Product Code. 11409516
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409516 1 g 1g
11419516 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409516 Supplier Thermo Scientific Alfa Aesar Supplier No. A19238.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1632-19-5 | C8H9NO6 | 215.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1632-19-5
Molecular Formula C8H9NO6
Molecular Weight (g/mol) 215.161
MDL Number MFCD00276587
InChI Key QCPBGHYMHYWRHG-UHFFFAOYSA-N
Synonym dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate, maybridge4_003392, acmc-20al76, 2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 2,5-dimethyl 3,4-dihydroxy-1h-pyrrole-2,5-dicarboxylate, methyl 3,4-dihydroxy-5-methoxycarbonyl pyrrole-2-carboxylate, 2z,5z-2,5-bis hydroxy methoxy methylidene pyrrolidine-3,4-dione, 1h-pyrrole-2,5-dicarboxylicacid, 3,4-dihydroxy-, 2,5-dimethyl ester
PubChem CID 2727733
IUPAC Name dimethyl 3,4-dihydroxy-1H-pyrrole-2,5-dicarboxylate
SMILES COC(=O)C1=C(C(=C(N1)C(=O)OC)O)O

Specifications

Melting Point 209°C to 212°C
Density 1.248
Boiling Point 268°C to 272°C
Flash Point >110°C (230°F)
Quantity 1 g
Formula Weight 215.16
Percent Purity 97%
Chemical Name or Material Dimethyl 3,4-dihydroxypyrrole-2,5-dicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.