missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetone-1,3-dicarboxylate, 97%

Product Code. 11407177
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11407177 25 g 25g
11417177 100 g 100g
11427177 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11407177 Supplier Thermo Scientific Alfa Aesar Supplier No. A14969.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 1830-54-2 | C7H10O5 | 174.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1830-54-2
Molecular Formula C7H10O5
Molecular Weight (g/mol) 174.15
MDL Number MFCD00008462
InChI Key RNJOKCPFLQMDEC-UHFFFAOYSA-N
Synonym dimethyl 1,3-acetonedicarboxylate, dimethyl acetone-1,3-dicarboxylate, dimethyl 3-oxoglutarate, 1,5-dimethyl 3-oxopentanedioate, pentanedioic acid, 3-oxo-, dimethyl ester, dimethyl acetonedicarboxylate, acetone dicarboxylic acid, dimethyl ester, glutaric acid, 3-oxo-, dimethyl ester, pentanedioic acid, 3-oxo-, 1,5-dimethyl ester, dimethyl 3-oxopentane-1,5-dioate
PubChem CID 74591
SMILES COC(=O)CC(=O)CC(=O)OC

Specifications

Melting Point 16°C to 17°C
Density 1.185
Boiling Point 237°C to 241°C
Flash Point >110°C (230°F)
Odor Ester-like
Refractive Index 1.444
Quantity 25 g
Beilstein 1366042
IUPAC Name 1,5-dimethyl 3-oxopentanedioate
Formula Weight 174.16
Percent Purity 97%
Chemical Name or Material Dimethyl acetone-1,3-dicarboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.