missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl acetylmethylphosphonate, 97%

Product Code. 11360094
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11360094 25 g 25g
11350094 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11360094 Supplier Thermo Scientific Alfa Aesar Supplier No. L00537.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4202-14-6 | C5H11O4P | 166.113 g/mol

Dimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl acetylmethylphosphonate is a Reagent used together with tosyl azide in a one-pot alkynylation of aldehydes.

Solubility
Difficult to mix.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4202-14-6
Molecular Formula C5H11O4P
Molecular Weight (g/mol) 166.113
MDL Number MFCD00008769
InChI Key UOWIYNWMROWVDG-UHFFFAOYSA-N
Synonym dimethyl 2-oxopropyl phosphonate, dimethyl acetylmethylphosphonate, dimethyl 2-oxopropylphosphonate, dimethyl acetonylphosphonate, 2-oxopropyl phosphonic acid dimethyl ester, 1-dimethoxycarbonyl acetone, acmc-209jmw, dimethoxyphosphoryl acetone
PubChem CID 77872
IUPAC Name 1-dimethoxyphosphorylpropan-2-one
SMILES CC(=O)CP(=O)(OC)OC

Specifications

Density 1.202
Boiling Point 121°C to 123°C (11 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.439
Quantity 25 g
Beilstein 1766218
Solubility Information Difficult to mix.
Formula Weight 166.12
Percent Purity 97%
Chemical Name or Material Dimethyl acetylmethylphosphonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.