missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl adipate, 99+%

Product Code. 10741461 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10741461

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-93-0 | C8H14O4 | 174.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-93-0
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.2
MDL Number MFCD00008469
InChI Key UDSFAEKRVUSQDD-UHFFFAOYSA-N
Synonym dimethyl adipate, dimethyladipate, adipic acid dimethyl ester, hexanedioic acid, dimethyl ester, adipic acid, dimethyl ester, 1,6-dimethylhexanedioate, methyl adipate van, 1,6-dimethyl hexanedioate, dbe 6, unii-by71rx0r62
PubChem CID 12329
ChEBI CHEBI:34715
IUPAC Name dimethyl hexanedioate
SMILES COC(=O)CCCCC(=O)OC

Specifications

Acidity 0.01% max. (as adipic acid)
Melting Point 8.0°C
Density 1.0600g/mL
Boiling Point 109.0°C to 110.0°C (14.0 mmHg)
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2C(CH2)4CO2CH3
Refractive Index 1.4270 to 1.4290
Quantity 100 g
Beilstein 02, 652
Merck Index 10, 151
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,alcohols,carbon tetrachloride,,acetic acid
Viscosity 5 mPa.s (20°C)
Formula Weight 174.2
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dimethyl adipate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl adipate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.