missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl adipimidate dihydrochloride, 97%

Product Code. 11338475
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338475 10 g 10g
11328475 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11338475 Supplier Thermo Scientific Alfa Aesar Supplier No. L10515.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14620-72-5 | C8H18Cl2N2O2 | 245.144 g/mol

Dimethyl adipimidate dihydrochloride was used as a cross-linking reagent in the study of rhodopsin in sheep photoreceptor membrane

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl adipimidate dihydrochloride was used as a cross-linking reagent in the study of rhodopsin in sheep photoreceptor membrane

Solubility
Hydrolyses in water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14620-72-5
Molecular Formula C8H18Cl2N2O2
Molecular Weight (g/mol) 245.144
MDL Number MFCD00012573
InChI Key IEUUDEWWMRQUDS-UHFFFAOYSA-N
Synonym dimethyl adipimidate dihydrochloride, unii-who0f998gm, who0f998gm, adipimidic acid dimethyl ester dihydrochloride, dimethyl adipimidate hcl, dmai, acmc-209tig, bicl300, dimethyladipimidate dihydrochloride
PubChem CID 12876176
IUPAC Name dimethyl hexanediimidate;dihydrochloride
SMILES COC(=N)CCCCC(=N)OC.Cl.Cl

Specifications

Melting Point ∽215°C (decomposition)
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Hydrolyses in water.
Formula Weight 245.15
Percent Purity 97%
Chemical Name or Material Dimethyl adipimidate dihydrochloride

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.