missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl allylmalonate, 97%

Product Code. 11317685
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317685 50 g 50g
11307685 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317685 Supplier Thermo Scientific Alfa Aesar Supplier No. L02376.18
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 40637-56-7 | C8H12O4 | 172.18 g/mol

Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl allylmalonate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Notes
Protect from moisture, heat. Incomaptible with oxidizing agents. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40637-56-7
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00014959
InChI Key VZNFVLWVVHHMBG-UHFFFAOYSA-N
Synonym dimethyl allylmalonate, dimethyl 2-allylmalonate, propanedioic acid, 2-propenyl-, dimethyl ester, 1,3-dimethyl 2-prop-2-en-1-yl propanedioate, dimethyl allyl-malonate, acmc-1bnbn, dimethyl 2-allylmalonate #, 2-allyl-propanedioic acid dimethyl ester, 2-prop-2-enylpropanedioic acid dimethyl ester
PubChem CID 574826
IUPAC Name dimethyl 2-prop-2-enylpropanedioate
SMILES COC(=O)C(CC=C)C(=O)OC

Spezifikation

Melting Point <-80°C
Density 1.074
Boiling Point 207°C
Flash Point 95°C (203°F)
Refractive Index 1.435
Quantity 50 g
Beilstein 1772822
Formula Weight 172.18
Percent Purity 97%
Chemical Name or Material Dimethyl allylmalonate

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.