missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl azelate, 97%

Product Code. p-7047051
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15409069

Brand: Thermo Scientific Alfa Aesar L17458.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1732-10-1 | C11H20O4 | 216.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1732-10-1
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00025898
InChI Key DRUKNYVQGHETPO-UHFFFAOYSA-N
Synonym dimethyl azelate, nonanedioic acid, dimethyl ester, methyl azelate, azelaic acid dimethyl ester, azelaic acid, dimethyl ester, 1,9-dimethyl nonanedioate, unii-29185k7oei, nonanedioic acid, 1,9-dimethyl ester, nonanedioic acid dimethyl ester, dimethyl nonane-1,9-dioate
PubChem CID 15612
SMILES COC(=O)CCCCCCCC(=O)OC

Specifications

Melting Point 18°C
Density 1.007
Boiling Point 156°C (20 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3OOC(CH2)7COOCH3
Refractive Index 1.437
Quantity 5 g
Beilstein 1710125
Merck Index 14,905
IUPAC Name 1,9-dimethyl nonanedioate
Formula Weight 216.28
Percent Purity 97%
Chemical Name or Material Dimethyl azelate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.