missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Carbonate, TRC

Product Code. 30376500
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
Product Code. Quantity unitSize
30376500 250 g 250g
1 options
This item is not returnable. View return policy

Product Code. 30376500

Brand: Toronto Research Chemicals TRCD446705250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl Carbonate
CAS 616-38-6
Molecular Formula C3 H6 O3
Quantity 250 g
Synonym Carbonic acid, dimethyl ester, Carbonic acid dimethyl ester, AK 100, AK 100 (carbonate), Dimethyl carbonate, Methyl carbonate, Methyl carbonate ((MeO)2CO), NSC 9371
InChI Formula InChI=1S/C3H6O3/c1-5-3(4)6-2/h1-2H3
SMILES COC(=O)OC
IUPAC Name dimethyl carbonate
Molecular Weight (g/mol) 90.0779
Formula Weight 90.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.