missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Chlorothiophosphate (>85%), TRC

Product Code. 30370630
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30370630 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370630 Supplier Toronto Research Chemicals Supplier No. TRCD473290500MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl chlorothiophosphate
CAS 2524-03-0
Molecular Formula C2 H6 Cl O2 P S
Quantity 500 mg
Synonym Phosphorochloridothioic Acid O,O-Dimethyl Ester, Methyl Phosphorochloridothioate ((MeO)2ClPS) (6CI,7CI), Chlorodimethoxyphosphine Sulfide, Dimethoxy Thiophosphonyl Chloride, Dimethyl Chlorophosphorothioate, Dimethyl Thionophosphorochloridate, Dimethyl Thiophosphorochloridate, NSC 132984, O,O-Dimethyl Chlorophosphorothioate, O,O-Dimethyl Chlorothionophosphate, O,O-Dimethyl Chlorothiophosphate, O,O-Dimethyl Phosphorochloridothioate, O,O-Dimethyl Phosphorochlorothioate, O,O-Dimethyl Phosphorothionochloridate, O,O-Dimethyl Thionophosphorochloridate, O,O-Dimethyl Thiophosphoric Acid Chloride, O,O-Dimethyl Thiophosphoryl Chloride, O,O-Dimethylthionophosphoryl Chloride
InChI Formula InChI=1S/C2H6ClO2PS/c1-4-6(3,7)5-2/h1-2H3
SMILES COP(=S)(Cl)OC
IUPAC Name chloro-dimethoxy-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 160.56
Formula Weight 159.9515
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.