missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cis-1,2-cyclopropanedicarboxylate, 97+%

Product Code. 10617041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10617041 5 g Glass bottle 5g
10195593 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10617041 Supplier Thermo Scientific Acros Supplier No. 255340050
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 826-34-6 | C7H10O4 | 158.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 826-34-6
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.16
MDL Number MFCD00062805
InChI Key JBVOSZYUSFDYIN-SYDPRGILSA-N
Synonym dimethyl cis-1,2-cyclopropanedicarboxylate, dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, cis-dimethyl cyclopropane-1,2-dicarboxylate, dimethylcis-1,2-cyclopropanedicarboxylate, cyclopropane-1,2-dicarboxylic acid dimethyl ester, z, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s-rel, 1,2-dimethyl 1r,2s-cyclopropane-1,2-dicarboxylate, 1r,2s-dimethyl cyclopropane-1,2-dicarboxylate, 1,2-cyclopropanedicarboxylic acid, dimethyl ester, 1r,2s, 1,2-cyclopropanedicarboxylicacid, 1,2-dimethyl ester, 1r,2s-rel
PubChem CID 642979
IUPAC Name dimethyl (1S,2R)-cyclopropane-1,2-dicarboxylate
SMILES COC(=O)C1CC1C(=O)OC

Specifications

Melting Point 34°C to 38°C
Color Colorless
Density 1.1500g/mL
Boiling Point 112.0°C (18.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4445 to 1.4465
Quantity 5 g
Specific Gravity 1.15
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in all organic solvents
Formula Weight 158.16
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Dimethyl cis-1,2-cyclopropanedicarboxylate
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl cis-1,2-cyclopropanedicarboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.