missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl fumarate, 99%

Product Code. 10206760 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10206760 25 g Glass bottle 25g
10591851 100 g Glass bottle 100g
10771725 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10206760 Supplier Thermo Scientific Acros Supplier No. 222180250
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 624-49-7 | C6H8O4 | 144.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-49-7
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.13
MDL Number MFCD00064438
InChI Key LDCRTTXIJACKKU-ONEGZZNKSA-N
Synonym dimethyl fumarate, tecfidera, e-dimethyl fumarate, fumaderm, dimethyl e-but-2-enedioate, fumaric acid, dimethyl ester, boletic acid dimethyl ester, fumaric acid dimethyl ester, methyl fumarate, dimethyl trans-ethylenedicarboxylate
PubChem CID 637568
ChEBI CHEBI:76004
IUPAC Name dimethyl (E)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point 102°C to 105°C
Color Colorless to Yellow
Boiling Point 192°C to 193°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2CCH=CHCO2CH3
Quantity 25 g
Beilstein 02, 741
Solubility Information Solubility in water: 1.6g/L (20°C)
Formula Weight 144.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl fumarate
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl fumarate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.