missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl glutarate, 98%

Product Code. 11425797
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425797 100 g 100g
11415797 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425797 Supplier Thermo Scientific Alfa Aesar Supplier No. B24359.22
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1119-40-0 | C7H12O4 | 160.17 g/mol

Dimethyl glutarate is applied as paint and graffiti removers, nail polish removers, hand cleaners, adhesives, sealants and caulks, automotive cleaning products, bathroom and tile cleaners, general purpose cleaners, spot removers, floor maintenance products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl glutarate is applied as paint and graffiti removers, nail polish removers, hand cleaners, adhesives, sealants and caulks, automotive cleaning products, bathroom and tile cleaners, general purpose cleaners, spot removers, floor maintenance products.

Solubility
Soluble in water. (53 g/L)

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-40-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00008468
InChI Key XTDYIOOONNVFMA-UHFFFAOYSA-N
Synonym dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester
PubChem CID 14242
SMILES COC(=O)CCCC(=O)OC

Specifications

Melting Point -37°C
Density 1.087
Boiling Point 213°C to 215°C
Flash Point 103°C (217°F)
Refractive Index 1.424
Quantity 100 g
Beilstein 1771444
Merck Index 14,4473
Solubility Information Soluble in water. (53g/L)
IUPAC Name dimethyl pentanedioate
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material Dimethyl glutarate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.