missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl glutarate, 98%

Product Code. 10192570 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10192570 500 g Glass bottle 500g
10182570 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10192570 Supplier Thermo Scientific Acros Supplier No. 116235000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1119-40-0 | C7H12O4 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-40-0
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00008468
InChI Key XTDYIOOONNVFMA-UHFFFAOYSA-N
Synonym dimethyl glutarate, glutaric acid dimethyl ester, pentanedioic acid, dimethyl ester, methyl glutarate, glutaric acid, dimethyl ester, dbe 5, unii-1i9vfa346p, pentanedioic acid, 1,5-dimethyl ester, 1,5-dimethyl pentanedioate, pentanedioic acid dimethyl ester
PubChem CID 14242
IUPAC Name dimethyl pentanedioate
SMILES COC(=O)CCCC(=O)OC

Specifications

Melting Point -37.0°C
Color Colorless
Density 1.0870g/mL
Boiling Point 210.0°C to 215.0°C
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3O2C(CH2)3CO2CH3
Refractive Index 1.4236 to 1.4256
Quantity 500 g
Beilstein 02, 633
Merck Index 15, 4509
Specific Gravity 1.087
Solubility Information Solubility in water: 53g/L
Viscosity 2.5 mm2/s (25°C)
Formula Weight 160.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dimethyl glutarate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.