missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl heptanebis(imidate) dihydrochloride, 98%, BLD Pharm™

Product Code. 31110334
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31110334 5 g
31109341 250 mg
31109190 1 g
31104050 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31110334 Supplier BLD Pharm Supplier No. BD1254575g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 58537-94-3
Molecular Formula C9H20Cl2N2O2
MDL Number MFCD00038942
Quantity 5 g
SMILES N=C(OC)CCCCCC(OC)=N.[H]Cl.[H]Cl
Molecular Weight (g/mol) 259.17
Percent Purity 98%
Chemical Name or Material Dimethyl heptanebis(imidate) dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.