missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Isophthalate, TRC

Product Code. 30378076
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378076 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30378076 Supplier Toronto Research Chemicals Supplier No. TRCD47404525G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isophthalic acid, bis-methyl ester
CAS 1459-93-4
Molecular Formula C10 H10 O4
Quantity 25 g
Synonym 1,3-Benzenedicarboxylic acid, 1,3-dimethyl ester, 1,3-Benzenedicarboxylic acid, dimethyl ester (9CI), Isophthalic acid, dimethyl ester (6CI,8CI), 1,3-Di(methoxycarbonyl)benzene, 1,3-Dimethyl benzene-1,3-dicarboxylate, DMIP, Dimethy isophthalate, Dimethyl 1,3-benzenedicarboxylate, Dimethyl isophthalate, Dimethyl m-phthalate, Methyl 3-carbomethoxybenzoate, Methyl isophthalate, Morflex 1129, NSC 15313, Isophthalic acid, bis-methyl ester, Isophthalic acid dimethyl ester
InChI Formula InChI=1S/C10H10O4/c1-13-9(11)7-4-3-5-8(6-7)10(12)14-2/h3-6H,1-2H3
SMILES COC(=O)c1cccc(c1)C(=O)OC
IUPAC Name dimethyl benzene-1,3-dicarboxylate
Molecular Weight (g/mol) 194.18
Formula Weight 194.0579
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.