missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl itaconate, 97%

Product Code. 11303296
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303296 100 g 100g
11313296 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303296 Supplier Thermo Scientific Alfa Aesar Supplier No. L12652.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 617-52-7 | C7H10O4 | 158.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-52-7
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00008446
InChI Key ZWWQRMFIZFPUAA-UHFFFAOYSA-N
Synonym dimethyl itaconate, dimethyl 2-methylenesuccinate, dimethylitaconate, itaconic acid dimethyl ester, itaconic acid, dimethyl ester, butanedioic acid, methylene-, dimethyl ester, 1,4-dimethyl 2-methylidenebutanedioate, dimethyl methylenesuccinate, unii-11jib0yi93, succinic acid, methylene-, dimethyl ester
PubChem CID 69240
IUPAC Name dimethyl 2-methylidenebutanedioate
SMILES COC(=O)CC(=C)C(=O)OC

Specifications

Melting Point 33°C to 39°C
Density 1.124
Boiling Point 207°C to 208°C
Flash Point 100°C (212°F)
Quantity 100 g
Beilstein 386674
Formula Weight 158.16
Percent Purity 97%
Chemical Name or Material Dimethyl itaconate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.