missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl maleate, 96%

Product Code. 11463188
Click to view available options
Quantity:
100 g
250 g
1000 g
5000 g
Unit Size:
1000g
100g
250g
5000g
This item is not returnable. View return policy

Product Code. 11463188

Brand: Thermo Scientific Alfa Aesar A18944.0I

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 624-48-6 | C6H8O4 | 144.126 g/mol

Dimethyl maleate is acts as a dienophile involved in ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. It is widely used as an intermediate and additive for pharmaceuticals, copolymers, pigments, plastics, paints, adhesives and agrochemicals. As an internal modifier, it increases the glass transition temperature of styrene or vinyl chloride polymer. It plays a vital role to improve the hardness and toughness of polymer films such as copolymers of vinyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl maleate is acts as a dienophile involved in ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. It is widely used as an intermediate and additive for pharmaceuticals, copolymers, pigments, plastics, paints, adhesives and agrochemicals. As an internal modifier, it increases the glass transition temperature of styrene or vinyl chloride polymer. It plays a vital role to improve the hardness and toughness of polymer films such as copolymers of vinyl acetate.

Solubility
Miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-48-6
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00008459
InChI Key LDCRTTXIJACKKU-ARJAWSKDSA-N
Synonym dimethyl maleate, maleic acid, dimethyl ester, methyl maleate, sipomer dmm, maleic acid dimethyl ester, dimethyl cis-butenedioate, 2-butenedioic acid z-, dimethyl ester, dimethyl cis-ethylenedicarboxylate, 2-butenedioic acid 2z-, dimethyl ester, 2-butenedioic acid, dimethyl ester, z
PubChem CID 5271565
ChEBI CHEBI:35460
IUPAC Name dimethyl (Z)-but-2-enedioate
SMILES COC(=O)C=CC(=O)OC

Specifications

Melting Point -19°C
Density 1.151
Boiling Point 204°C to 205°C
Flash Point 91°C (195°F)
Refractive Index 1.442
Quantity 5000 g
UN Number UN2810
Beilstein 471705
Solubility Information Miscible with water.
Formula Weight 144.13
Percent Purity 96%
Chemical Name or Material Dimethyl maleate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.