missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl malonate, 98+%

Product Code. 11414473
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414473 100 g 100g
11424473 500 g 500g
11434473 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11414473 Supplier Thermo Scientific Alfa Aesar Supplier No. A11007.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-59-8 | C5H8O4 | 132.115 g/mol

Dimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl malonate is a reagent used in organic synthesis. It acts as a precursor for the synthesis of mono-substituted and di-substituted acetic acid, barbiturates, vitamin B1 and vitamin B6. It is used in the pharmaceutical industry to prepare pharmaceuticals like chloroquine and butazolidin. It is involved in the synthesis of diastereomeric pure thienylpyridines by reacting with 1,3-diphenylprop-2-enyl acetate.

Solubility
Miscible with alcohol, ether and oils. Slightly miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-59-8
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.115
MDL Number MFCD00008460
InChI Key BEPAFCGSDWSTEL-UHFFFAOYSA-N
Synonym dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
PubChem CID 7943
IUPAC Name dimethyl propanedioate
SMILES COC(=O)CC(=O)OC

Specifications

Melting Point -62°C
Density 1.156
Boiling Point 180°C to 181°C
Flash Point 90°C (194°F)
Refractive Index 1.413
Quantity 100 g
Beilstein 774261
Merck Index 14,6096
Solubility Information Miscible with alcohol,ether and oils. Slightly miscible with water.
Formula Weight 132.12
Percent Purity ≥98%
Chemical Name or Material Dimethyl malonate
Show More Show Less
compliance-icons
Product Identifier
  • Dimethyl malonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Dimethyl malonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.