missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl malonate, 99+%

Product Code. 13448478 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13448478 5 L Plastic drum 5 L
10657362 250 mL Glass bottle 250mL
10647362 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 13448478 Supplier Thermo Scientific Acros Supplier No. 220700051
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-59-8 | C5H8O4 | 132.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-59-8
Molecular Formula C5H8O4
Molecular Weight (g/mol) 132.12
MDL Number MFCD00008460
InChI Key BEPAFCGSDWSTEL-UHFFFAOYSA-N
Synonym dimethyl malonate, methyl malonate, propanedioic acid, dimethyl ester, malonic acid dimethyl ester, 1,3-dimethyl propanedioate, dimethyl 1,3-propanedioate, malonic acid, dimethyl ester, unii-em8y79998c, ccris 8981, propanedioic acid dimethyl ester
PubChem CID 7943
IUPAC Name dimethyl propanedioate
SMILES COC(=O)CC(=O)OC

Specifications

Melting Point -62.0°C
Color Colorless
Density 1.1560g/mL
Boiling Point 180.0°C to 181.0°C
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic drum
Linear Formula CH3O2CCH2CO2CH3
Refractive Index 1.4120 to 1.4140
Quantity 5 L
Beilstein 02,572
Fieser 08,193; 13,34
Merck Index 15,6168
Specific Gravity 1.156
Solubility Information Solubility in water: negligible. Other solubilities: soluble in alcohol and ether
Formula Weight 132.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dimethyl malonate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.