missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl N-Cyanodithioiminocarbonate, TRC

Product Code. 30383109
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383109 25 g
30369585 10 g
30369261 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383109 Supplier Toronto Research Chemicals Supplier No. TRCD47200025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 10191-60-3
Molecular Formula C4H6N2S2
Molecular Weight (g/mol) 146.23
Synonym (Cyanimino)bis(methylthio)methane, (Cyanimino)dimethyldithiocarbonate, Cyanocarbonodithioimidic acid dimethyl ester, Dimethyl (N-cyano) carbonimidodithioate, Dimethyl N-cyanodithiocarbonimidate, Dimethyl N-cyanodithioimidocarbonate, Dimethyl N-cyanodithioiminocarbonate, Dimethyl N-cyanoimidodithiocarbonate, Dimethyl N-cyanoiminodithiocarbonate, Dimethyl cyanimidodithiocarbonate, Dimethyl cyanocarbonimidodithioate, Dimethyl cyanodithioimidocarbonate, Dimethyl cyanoimidodithiocarbonate, Dimethyl cyanoiminodithiocarbonate, N-Cyanoiminodithiocarbonic acid dimethyl ester, N-[Bis(methylthio)methylene]cyanamide, NSC 145987, S,S-Dimethyl (N-cyanoimido)dithiocarbonate, S,S-Dimethyl cyanoimidodithiocarbonate, S,S-Dimethyl cyanoiminodithiocarbamate, S,S'-Dimethyl N-cyanodithioiminocarbonate, S,S'-Dimethyl cyanodithioimidocarbonate
IUPAC Name bis(methylsulfanyl)methylidenecyanamide
SMILES CSC(=NC#N)SC

Tekniske data

Quantity 25 g
InChI Formula InChI=1S/C4H6N2S2/c1-7-4(8-2)6-3-5/h1-2H3
Formula Weight 145.9972
Chemical Name or Material Dimethyl N-Cyanodithioiminocarbonate
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.