missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl oxalate, 99%

Product Code. 10406820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10406820 500 g Plastic bottle 500g
10347670 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10406820 Supplier Thermo Scientific Acros Supplier No. 159275000
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 553-90-2 | C4H6O4 | 118.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-90-2
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00008442
InChI Key LOMVENUNSWAXEN-UHFFFAOYSA-N
Synonym methyl oxalate, ethanedioic acid, dimethyl ester, oxalic acid dimethyl ester, dimethyloxalate, oxalic acid, dimethyl ester, dimethyl ethanedioate, unii-iq3q79344s, ethanedioic acid, 1,2-dimethyl ester, dimethyl ethane-1,2-dioate, dimethyl ester of oxalic acid
PubChem CID 11120
ChEBI CHEBI:6859
IUPAC Name dimethyl oxalate
SMILES COC(=O)C(=O)OC

Specifications

Melting Point 51°C to 55°C
Color Colorless to White
Density 1.1400g/mL
Boiling Point 163°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3O2CCO2CH3
Quantity 500 g
Beilstein 02, 534
Merck Index 15, 6179
Specific Gravity 1.14
Solubility Information Solubility in water: 60g/L (25°C). Other solubilities: soluble in ethanol,ether,acetone,chloroform
Formula Weight 118.09
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Dimethyl oxalate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.