missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl phenylphosphonite, 98%

Product Code. 11384479
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384479 1 g 1g
11467866 5 g 5g
11477866 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384479 Supplier Thermo Scientific Alfa Aesar Supplier No. A14006.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2946-61-4 | C8H11O2P | 170.15 g/mol

Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl phenylphosphonite is used in the preparation of phenyl-phosphonic acid dimethyl ester. It is used as a precursor for the preparation of dinitrosyltris(dimethyl phenylphosphonite)manganese(I) tetrafluoroborate.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2946-61-4
Molecular Formula C8H11O2P
Molecular Weight (g/mol) 170.15
MDL Number MFCD00008352
InChI Key LMZLQYYLELWCCW-UHFFFAOYSA-N
Synonym dimethyl phenylphosphonite, dimethoxyphenylphosphine, phenyldimethoxyphosphine, dimethylphenylphosphonite, phosphorus, dimethoxy phenyl, phosphonous acid, phenyl-, dimethyl ester, phenylphosphonous acid dimethyl ester, acmc-20al6n, php ome 2, dimethoxy phenyl phosphane
PubChem CID 76277
IUPAC Name dimethoxy(phenyl)phosphane
SMILES COP(OC)C1=CC=CC=C1

Specifications

Density 1.072
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 1 g
Beilstein 1073018
Sensitivity Air sensitive
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 170.15
Percent Purity 98%
Chemical Name or Material Dimethyl phenylphosphonite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.