missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl phosphate, 98%

Product Code. 10490731 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10490731

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 813-78-5 | C2H7O4P | 126.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 813-78-5
Color Brown or Colorless to Yellow
Density 1.3230g/mL
Boiling Point 174°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C2H7O4P
Refractive Index 1.4080 to 1.41
MDL Number MFCD00014887
Quantity 1 g
Beilstein 01,III,1205
Specific Gravity 1.323
Synonym dimethyl phosphate, dimethylphosphate, phosphoric acid, dimethyl ester, o,o-dimethyl phosphate, unii-hi4k2c9uei, methyl phosphate meo 2 ho po, o,o-dimethyl hydrogen phosphate, hi4k2c9uei, dimethoxyphosphinic acid, dimethyl acid phosphate
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in ethanol,acetone
InChI Key KKUKTXOBAWVSHC-UHFFFAOYSA-N
SMILES COP(=O)(O)OC
IUPAC Name dimethyl hydrogen phosphate
Molecular Weight (g/mol) 126.02
PubChem CID 13134
Formula Weight 126.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Dimethyl phosphate, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl phosphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Phosphoric acid, dimethyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.