missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Phthalate, 99%

Product Code. 10367673 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
250mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367673 2.5 L Glass bottle 2.5 L
10306700 250 mL Glass bottle 250mL
10477680 1 L Glass bottle 1 L
10657953 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10367673 Supplier Thermo Scientific Supplier No. 116640025
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 131-11-3 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-11-3
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008425
InChI Key NIQCNGHVCWTJSM-UHFFFAOYSA-N
Synonym dimethyl phthalate, dimethylphthalate, solvanom, solvarone, avolin, fermine, phthalic acid dimethyl ester, mipax, palatinol m, unimoll dm
PubChem CID 8554
ChEBI CHEBI:4609
IUPAC Name dimethyl benzene-1,2-dicarboxylate
SMILES COC(=O)C1=CC=CC=C1C(=O)OC

Specifications

Density 1.1900g/mL
Boiling Point 282°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5140 to 1.516
Quantity 2.5 L
Beilstein 09, 797
Merck Index 15, 3279
Specific Gravity 1.19
Solubility Information Solubility in water: negligible. Other solubilities: miscible with organic solvents,insoluble in paraffinic hydrocarbons,slightly soluble in mineral oil
Viscosity 17 mPa.s (20°C)
Formula Weight 194.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl phthalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.