missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Phthalate, TRC

Product Code. 30135696
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135696 100 g 100g
17988124 10 g 10g
17968424 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135696 Supplier Toronto Research Chemicals Supplier No. TRCD477975100G
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-11-3
Molecular Formula C10 H10 O4
Molecular Weight (g/mol) 194.18
Synonym 1,2-Benzenedicarboxylic acid, 1,2-dimethyl ester, 1,2-Benzenedicarboxylic acid, dimethyl ester (9CI), Phthalic acid, dimethyl ester (6CI,8CI), 1,2-Dimethyl benzene-1,2-dicarboxylate, Avolin, DMF (insect repellent), DMP, Dimethyl 1,2-benzenedicarboxylate, Dimethyl o-phthalate, Dimethyl phthalate, Fermine, Metox 50, Mipax, NSC 15398, NTM, Palatinol M, Repeftal, Solvanom, Solvarone, Unimoll DM
IUPAC Name dimethyl benzene-1,2-dicarboxylate
SMILES COC(=O)c1ccccc1C(=O)OC

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6H,1-2H3
Formula Weight 194.0579
Chemical Name or Material Phthalic acid, bis-methyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.