missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl selenide

Product Code. 11364789
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11364789

Brand: Thermo Scientific Alfa Aesar 068116.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 593-79-3 | C2H6Se | 109.041 g/mol

Used as primary and secondary intermediate and also used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-79-3
Molecular Formula C2H6Se
Molecular Weight (g/mol) 109.041
MDL Number MFCD00014848
InChI Key RVIXKDRPFPUUOO-UHFFFAOYSA-N
Synonym dimethylselenide, dimethylselenium, dimethyl selenide, methyl selenide, methyl selenium, methane, selenobis, selenium dimethyl, selenide, dimethyl, ch3 2se, unii-yk0r6jkt6h
PubChem CID 11648
ChEBI CHEBI:4610
IUPAC Name methylselanylmethane
SMILES C[Se]C

Specifications

Color Colorless
Density 1.41 g/mL
Boiling Point 56°C to 58°C
Odor Unpleasant
Linear Formula (CH3)2Se
Quantity 5 g
UN Number UN3282
Beilstein 1696848
Sensitivity Air and light sensitive
Solubility Information Very soluble in ethyl ether and ethyl alcohol.
Formula Weight 109.03
Physical Form Liquid
Chemical Name or Material Dimethyl selenide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.