missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl suberate, 99%, ACROS Organics™

Product Code. 10347390 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10347390

Brand: Thermo Scientific Acros 165040250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1732-09-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00008471
InChI Key LNLCRJXCNQABMV-UHFFFAOYSA-N
Synonym dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate
PubChem CID 15611
ChEBI CHEBI:81345
IUPAC Name 1,8-dimethyl octanedioate
SMILES COC(=O)CCCCCCC(=O)OC

Specifications

Color Colorless to Yellow
Density 1.01g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3O2C(CH2)6CO2CH3
Refractive Index 1.4325 to 1.4345 (20°C, 589nm)
Quantity 25 g
Beilstein 02, 693
Formula Weight 202.25
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Dimethyl suberate, 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.