missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl suberate, 99%, ACROS Organics™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.0100g/mL |
| Boiling Point | 268.0°C |
| Flash Point | >110°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Linear Formula | CH3O2C(CH2)6CO2CH3 |
| Refractive Index | 1.4325 to 1.4345 |
| Quantity | 500 g |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction