missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl suberate, 99%

Product Code. 11485317
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11485317

Brand: Thermo Scientific Alfa Aesar B22681.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1732-09-8 | C10H18O4 | 202.25 g/mol

Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1732-09-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00008471
InChI Key LNLCRJXCNQABMV-UHFFFAOYSA-N
Synonym dimethyl suberate, octanedioic acid, dimethyl ester, suberic acid, dimethyl ester, octanedioic acid dimethyl ester, suberic acid dimethyl ester, octanedioic acid dimethyl, 1,8-dimethyl octanedioate, octanedioic acid, 1,8-dimethyl ester, dimethyl octane-1,8-dioate
PubChem CID 15611
ChEBI CHEBI:81345
IUPAC Name dimethyl octanedioate
SMILES COC(=O)CCCCCCC(=O)OC

Specifications

Density 1.016
Boiling Point 266°C to 268°C
Flash Point >110°C (230°F)
Refractive Index 1.432
Quantity 25 g
Beilstein 1780054
Formula Weight 202.25
Percent Purity 99%
Chemical Name or Material Dimethyl suberate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.