missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl suberate, 99%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Dimethyl suberate was used in the preparation of [8-2H2] and [7-2H2]oleates.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 1.016 |
| Boiling Point | 266°C to 268°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.432 |
| Quantity | 25 g |
| Beilstein | 1780054 |
| Formula Weight | 202.25 |
| Percent Purity | 99% |
| Chemical Name or Material | Dimethyl suberate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction