missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl succinate, 98%

Product Code. 11364479
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
This item is not returnable. View return policy

Product Code. 11364479

Brand: Thermo Scientific Alfa Aesar A12565.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 106-65-0 | C6H10O4 | 146.142 g/mol

Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl succinate is used as a flavoring agent. It have wide range of industrial applications such as Functional fluids (open systems), Intermediates, Paint additives and coating additives, Pigments Solvents, Viscosity adjustors.

Solubility
Soluble in water. (122,9 g/L at 20°C). soluble in acetone, ethanol, very soluble in ethyl ether.

Notes
Store at room temperature. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-65-0
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.142
MDL Number MFCD00008466
InChI Key MUXOBHXGJLMRAB-UHFFFAOYSA-N
Synonym dimethyl succinate, methyl succinate, dimethylsuccinate, butanedioic acid, dimethyl ester, succinic acid, dimethyl ester, dbe-4 dibasic ester, methyl butanedioate, succinic acid dimethyl ester, butanedioic acid, 1,4-dimethyl ester, fema no. 2396
PubChem CID 7820
IUPAC Name dimethyl butanedioate
SMILES COC(=O)CCC(=O)OC

Specifications

Melting Point 18°C to 19°C
Density 1.119
Boiling Point 196°C
Flash Point 85°C (185°F)
Refractive Index 1.419
Quantity 100 g
Beilstein 956776
Merck Index 14,8869
Solubility Information Soluble in water. (122,9g/L at 20°C). soluble in acetone,ethanol,very soluble in ethyl ether.
Formula Weight 146.14
Percent Purity ≥98%
Chemical Name or Material Dimethyl succinate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.