missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl succinate, 99%

Product Code. 10736551 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10736551 100 g Glass bottle 100g
10193530 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10736551 Supplier Thermo Scientific Acros Supplier No. 150731000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 106-65-0 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-65-0
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00008466
InChI Key MUXOBHXGJLMRAB-UHFFFAOYSA-N
Synonym dimethyl succinate, methyl succinate, dimethylsuccinate, butanedioic acid, dimethyl ester, succinic acid, dimethyl ester, dbe-4 dibasic ester, methyl butanedioate, succinic acid dimethyl ester, butanedioic acid, 1,4-dimethyl ester, fema no. 2396
PubChem CID 7820
IUPAC Name dimethyl butanedioate
SMILES COC(=O)CCC(=O)OC

Specifications

Acidity 0.2mg KOH/g max.
Melting Point 18.0°C to 19.0°C
Density 1.1100g/mL
Boiling Point 200.0°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2CCH2CH2CO2CH3
Refractive Index 1.4185 to 1.4205
Quantity 100 g
Beilstein 02, 609
Merck Index 15, 8999
Specific Gravity 1.11
Solubility Information Solubility in water: 75g/L (20°C)
Formula Weight 146.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Dimethyl succinate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.