missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfide, 99+%

Product Code. 11308227
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11308227 1 L 1 L
11398217 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308227 Supplier Thermo Scientific Alfa Aesar Supplier No. 022949.K2
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-18-3 | C2H6S | 62.13 g/mol

Dimethyl sulfide is used as a displaceable ligand in chloro(dimethyl sulfide)gold(I) and other coordination compounds, as a reducing agent in ozonolysis reactions as well as a food flavoring component. Further, it is utilized for the preparation of dimethyl sulfoxide, which is a common and important industrial solvent. It finds application in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-18-3
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00008562
InChI Key QMMFVYPAHWMCMS-UHFFFAOYSA-N
Synonym dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane
PubChem CID 1068
ChEBI CHEBI:17437
SMILES CSC

Specifications

Melting Point -83°C
Density 0.845 g/mL
Boiling Point 38°C
Flash Point -48°C (-54°F)
Odor Unpleasant
Refractive Index 1.4351
Quantity 1 L
UN Number UN1164
Merck Index 14,6123
Solubility Information Miscible with alcohols,ethers,esters,ketones,aliphatic and aromatic hydrocarbons. Slightly miscible with
IUPAC Name methylsulfanylmethane
Formula Weight 62.14
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Dimethyl sulfide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethyl sulfide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Dimethyl sulfide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.