missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfide, TRC

Product Code. 30450163
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450163 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450163 Supplier Toronto Research Chemicals Supplier No. TRCT3480001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl sulfide
CAS 75-18-3
Molecular Formula C2 H6 S
Quantity 1 g
Synonym Methane','thiobis- (9CI), Methyl sulfide (8CI), 1,1'-Thiobis[methane], (Methylsulfanyl)methane, 2-Thiapropane, DMS, Dimethyl monosulfide, Dimethyl sulfide, Dimethyl sulphide, Dimethyl thioether, Dimethylsulfane, Methyl monosulfide, Methylthiomethane, Thiobis(methane), 1,1'-Thiobismethane
InChI Formula InChI=1S/C2H6S/c1-3-2/h1-2H3
SMILES CSC
IUPAC Name methylsulfanylmethane
Molecular Weight (g/mol) 62.13
Formula Weight 62.019
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.